Assay Details
Target Gene Details
Entrez Gene ID: | 77626 |
Gene Name: | sphingomyelin phosphodiesterase 4 |
Gene Aliases: |
4122402O22Rik, mKIAA1418 |
Location: |
Chr.16:17619354-17644830 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smpd4 | NM_001164609.1 | NP_001158081.1 | ||
NM_001164610.1 | NP_001158082.1 | |||
NM_001164611.1 | NP_001158083.1 | |||
NM_029945.3 | NP_084221.2 | |||
AK019466.2 | BAB31737.1 | |||
AK031029.1 | BAC27221.1 | |||
AK031685.1 | BAC27512.1 | |||
AK035892.1 | BAC29229.1 | |||
AK036055.1 | ||||
AK036380.1 | ||||
AK129354.1 | BAC98164.1 | |||
AK138720.1 | ||||
AK164234.1 | BAE37694.1 | |||
AK164456.1 | BAE37796.1 | |||
AK179025.1 | ||||
AK189409.1 | ||||
AK191437.1 | ||||
AK196995.1 | ||||
BC026767.1 | AAH26767.1 |
More Information
Set Membership: |
Intragenic Non-exonic |