Assay Details
Target Gene Details
Entrez Gene ID: | 15557 |
Gene Name: | 5-hydroxytryptamine (serotonin) receptor 1F |
Gene Aliases: |
Htr1eb |
Location: |
Chr.16:64922423-65105784 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Htr1f | NM_008310.3 | NP_032336.1 | ||
AK045364.1 | BAC32328.1 | |||
BC120507.1 | ||||
BC137604.1 | ||||
Z14224.1 | CAA78593.1 |
More Information
Set Membership: |
Intragenic Non-exonic |