Assay Details
Target Gene Details
Entrez Gene ID: | 224020 |
Gene Name: | phosphatidylinositol 4-kinase, catalytic, alpha polypeptide |
Gene Aliases: |
Pik4ca |
Location: |
Chr.16:17280351-17406314 on Build GRCm38 |
Assay Gene Location: | Within Exon 39 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pi4ka | NM_001001983.2 | 39 | 4492 | NP_001001983.2 |
AK136942.1 | 3 | 279 | BAE23184.1 | |
BC075629.1 | 39 | 4492 | AAH75629.1 |
More Information
Set Membership: |
Intragenic Exonic |