Assay Details
Target Gene Details
Entrez Gene ID: | 78913 |
Gene Name: | listerin E3 ubiquitin protein ligase 1 |
Gene Aliases: |
4930528H02Rik, AV266914, C87237, Listerin, Rnf160, Zfp294 |
Location: |
Chr.16:87376651-87432619 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ltn1 | NM_001081068.1 | 12 | 2440 | NP_001074537.1 |
AK036725.1 | 12 | 2370 | BAC29552.1 | |
AK165062.1 | 12 | 2360 | ||
AK173009.1 | 12 | 2311 | BAD32287.1 |
More Information
Set Membership: |
Intragenic Exonic |