Assay Details
Target Gene Details
Entrez Gene ID: | 224092 |
Gene Name: | large subunit GTPase 1 homolog (S. cerevisiae) |
Gene Aliases: |
5830465I20, AA409273, D16Bwg1547e |
Location: |
Chr.16:30561369-30587589 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lsg1 | NM_178069.5 | 7 | 727 | NP_835170.1 |
AK030940.1 | 7 | 702 | ||
AK078501.1 | 7 | 700 | ||
AK132527.1 | 8 | 930 | ||
AK145186.1 | 7 | 702 | BAE26280.1 | |
AK146895.1 | 7 | 702 | BAE27512.1 | |
AK186672.1 | ||||
BC043724.1 | 7 | 727 | AAH43724.1 | |
BC080306.1 | 5 | 458 | AAH80306.1 |