Assay Details
Target Gene Details
Entrez Gene ID: | 16419 |
Gene Name: | integrin beta 5 |
Gene Aliases: |
AA475909, AI874634, ESTM23, [b]-5, [b]5, [b]5A, [b]5B, beta-5, beta5 |
Location: |
Chr.16:33829665-33949338 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb5 | NM_001145884.1 | NP_001139356.1 | ||
NM_010580.2 | NP_034710.2 | |||
AF022110.1 | AAD08782.1 | |||
AF043256.1 | AAC40109.1 | |||
AF043257.1 | AAC40110.1 | |||
AK087476.1 | ||||
AK140251.1 | BAE24299.1 | |||
AK143532.1 | ||||
BC046280.1 | ||||
BC058246.1 | AAH58246.1 |
More Information
Set Membership: |
Intragenic Non-exonic |