Assay Details
Target Gene Details
Entrez Gene ID: | 15233 |
Gene Name: | homogentisate 1, 2-dioxygenase |
Gene Aliases: |
Hgo, aku |
Location: |
Chr.16:37580153-37632026 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hgd | NM_013547.3 | 9 | 767 | NP_038575.2 |
NR_027778.1 | 8 | 678 | ||
AK002584.1 | 8 | 638 | ||
BC024369.1 | 8 | 581 | ||
BC037628.1 | 9 | 681 | AAH37628.2 | |
BC044758.2 | 8 | 567 | ||
BC055029.1 | 9 | 648 | AAH55029.1 |
More Information
Set Membership: |
Intragenic Exonic |