Assay Details
Target Gene Details
Entrez Gene ID: | 94184 |
Gene Name: | pyridoxal-dependent decarboxylase domain containing 1 |
Gene Aliases: |
2210010A19Rik, AA415817, Kiaa0251-hp |
Location: |
Chr.16:13833161-13903145 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdxdc1 | NM_001039533.2 | 6 | 703 | NP_001034622.1 |
NM_001291017.1 | 6 | 703 | NP_001277946.1 | |
NM_053181.3 | 6 | 703 | NP_444411.2 | |
AK004611.1 | 6 | 580 | BAB23408.1 | |
AK030613.1 | 6 | 534 | BAC27047.1 | |
AK033107.1 | 6 | 534 | BAC28155.1 | |
AK054329.1 | 6 | 686 | BAC35732.1 | |
AK163696.1 | 6 | 574 | BAE37462.1 | |
AK165587.1 | 6 | 538 | BAE38276.1 | |
AK170661.1 | 6 | 690 | BAE41943.1 | |
BC005541.1 | 6 | 521 | AAH05541.1 |