Assay Details
Target Gene Details
Entrez Gene ID: | 74197 |
Gene Name: | general transcription factor II E, polypeptide 1 (alpha subunit) |
Gene Aliases: |
2610024P03Rik, 56kDa, AI303800, FE, TFIIE-A |
Location: |
Chr.16:37509790-37539769 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf2e1 | NM_028812.3 | NP_083088.1 | ||
AK011543.1 | BAB27686.1 | |||
AK031528.1 | BAC27436.1 | |||
AK080478.1 | BAC37928.1 | |||
AK154283.1 | BAE32487.1 | |||
BC056966.1 | AAH56966.1 |
More Information
Set Membership: |
Intragenic Non-exonic |