Assay Details
Target Gene Details
Entrez Gene ID: | 21832 |
Gene Name: | thrombopoietin |
Gene Aliases: |
Mgdf, Ml, Mpllg, Tpo |
Location: |
Chr.16:20724454-20734511 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Thpo | NM_001173505.1 | 2 | 209 | NP_001166976.1 |
NM_001289894.1 | 2 | 209 | NP_001276823.1 | |
NM_001289896.1 | 2 | 209 | NP_001276825.1 | |
NM_009379.3 | 2 | 363 | NP_033405.1 | |
AI314551.1 | 2 | 209 | ||
AK047512.1 | 2 | 364 | BAC33078.1 | |
BC003803.1 | 2 | 475 | AAH03803.1 | |
L34169.1 | 2 | 146 | AAA40436.1 |