Assay Details
Target Gene Details
Entrez Gene ID: | 106298 |
Gene Name: | RRN3 RNA polymerase I transcription factor homolog (yeast) |
Gene Aliases: |
AL023001, E130302O19Rik, R75565, TIF-1A, Tif1a |
Location: |
Chr.16:13780699-13814841 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rrn3 | NM_001039521.1 | 12 | 1105 | NP_001034610.1 |
AK080397.1 | 12 | 1061 | ||
AK165676.1 | 12 | 1094 | ||
BC034110.1 | 8 | 620 | AAH34110.1 | |
BC138774.1 | 12 | 1070 | ||
BC138776.1 | 12 | 1070 |
More Information
Set Membership: |
Intragenic Exonic |