Assay Details
Target Gene Details
Entrez Gene ID: | 320355 |
Gene Name: | lipase, member I |
Gene Aliases: |
D930038D03Rik, lpd1 |
Location: |
Chr.16:75540514-75586072 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lipi | NM_001252513.1 | 9 | 1217 | NP_001239442.1 |
AK079027.1 | 7 | 628 | BAC37507.1 | |
AK086576.1 | 9 | 1222 | ||
BC147403.1 | 4 | 409 | ||
BC147404.1 | 4 | 409 |
More Information
Set Membership: |
Intragenic Exonic |