Assay Details
Target Gene Details
Entrez Gene ID: | 13845 |
Gene Name: | Eph receptor B3 |
Gene Aliases: |
AW456895, Cek10, Etk2, HEK2, MDK5, Sek4, Tyro6 |
Location: |
Chr.16:21204795-21223309 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ephb3 | NM_010143.1 | 8 | 2053 | NP_034273.1 |
AK144414.1 | 8 | 2069 | ||
AK158043.1 | 9 | 2993 | BAE34333.1 | |
BC014822.1 | 8 | 2054 | AAH14822.1 | |
BC053085.1 | 8 | 2053 | AAH53085.1 | |
U11493.1 | 6 | 1513 | AAA67925.1 | |
Z49086.1 | 8 | 1911 | CAA88910.1 |
More Information
Set Membership: |
Intragenic Exonic |