Assay Details
Target Gene Details
Entrez Gene ID: | 268878 |
Gene Name: | ATPase type 13A5 |
Gene Aliases: |
C630015F21Rik |
Location: |
Chr.16:29231851-29378764 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp13a5 | NM_001284375.1 | 11 | 1436 | NP_001271304.1 |
NM_175650.4 | 12 | 1571 | NP_783581.2 | |
AK083102.1 | 12 | 1546 | ||
AK083121.1 | 12 | 1572 | BAC38770.1 | |
AK158355.1 | 12 | 1391 | BAE34470.1 | |
BC115739.1 | 11 | 1369 |