Assay Details
Target Gene Details
Entrez Gene ID: | 268878 |
Gene Name: | ATPase type 13A5 |
Gene Aliases: |
C630015F21Rik |
Location: |
Chr.16:29231851-29378764 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp13a5 | NM_001284375.1 | 10 | 1222 | NP_001271304.1 |
NM_175650.4 | 11 | 1357 | NP_783581.2 | |
AK083102.1 | 11 | 1332 | ||
AK083121.1 | 11 | 1358 | BAC38770.1 | |
AK158355.1 | 11 | 1177 | BAE34470.1 | |
BC115739.1 | 10 | 1155 |