Assay Details
Target Gene Details
Entrez Gene ID: | 100038602 |
Gene Name: | predicted gene 10865 |
Gene Aliases: |
ENSMUSG00000075558 |
Location: |
Chr.15:78994955-79011041 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm10865 | NR_045746.1 | |||
AK139534.1 | BAE24054.1 |
Target Gene Details
Entrez Gene ID: | 110253 |
Gene Name: | TRIO and F-actin binding protein |
Gene Aliases: |
AI428493, EST478828, Tara, mKIAA1662 |
Location: |
Chr.15:78947724-79005869 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Triobp | NM_001024716.1 | NP_001019887.1 | ||
NM_001039156.1 | NP_001034245.1 | |||
NM_138579.4 | NP_613045.3 | |||
AK053194.1 | BAC35308.1 | |||
AK085512.1 | ||||
AK129417.1 | BAC98227.2 | |||
AK155197.1 | BAE33111.1 | |||
AK170390.1 | BAE41764.1 | |||
AK178072.1 | ||||
AK186966.1 | ||||
BC003984.1 | AAH03984.1 | |||
BC042760.1 | AAH42760.1 | |||
BC053329.1 | AAH53329.1 | |||
BU058377.1 | ||||
CA751843.1 | ||||
DQ228000.1 | ABB59556.1 | |||
DQ228001.2 | ABB59557.2 | |||
DQ278602.1 | ABB77203.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |