Assay Details
Target Gene Details
Entrez Gene ID: | 29813 |
Gene Name: | zinc finger protein 385A |
Gene Aliases: |
Hzf, Zfp385, Znf385a |
Location: |
Chr.15:103313895-103340086 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp385a | NM_013866.2 | NP_038894.2 | ||
AF118566.1 | AAF24093.1 | |||
AK137748.1 | ||||
BC017644.1 | AAH17644.1 |