Assay Details
Target Gene Details
Entrez Gene ID: | 223435 |
Gene Name: | triple functional domain (PTPRF interacting) |
Gene Aliases: |
6720464I07Rik, AA408740, Solo |
Location: |
Chr.15:27730649-28025953 on Build GRCm38 |
Assay Gene Location: | Within Exon 47 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trio | NM_001081302.1 | 47 | 7059 | NP_001074771.1 |
BC051169.1 | 6 | 526 | AAH51169.1 |
More Information
Set Membership: |
Intragenic Exonic |