Assay Details
Target Gene Details
Entrez Gene ID: | 75002 |
Gene Name: | transmembrane (C-terminal) protease, serine 12 |
Gene Aliases: |
4930478A21Rik |
Location: |
Chr.15:100280837-100293053 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmprss12 | NM_183109.3 | 2 | 246 | NP_898932.2 |
AK015583.1 | 2 | 259 | BAC25466.1 | |
AK132967.1 | 2 | 247 | BAE21447.1 |
More Information
Set Membership: |
Intragenic Exonic |