Assay Details
Target Gene Details
Entrez Gene ID: | 14167 |
Gene Name: | fibroblast growth factor 12 |
Gene Aliases: |
AV114868, B230343J05Rik, FGF-12, FHF-1, Fhf1 |
Location: |
Chr.16:28157783-28753243 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgf12 | NM_001276420.1 | 2 | 1187 | NP_001263349.1 |
AF020738.1 | AAB71607.1 | |||
AF020739.1 | AAB71608.1 | |||
AK011712.2 | ||||
AK018774.1 | 3 | 327 | ||
AK034335.1 | BAC28679.1 | |||
AK148215.1 | 2 | 1188 | BAE28417.1 | |
BB611389.1 | ||||
BC030485.1 | AAH30485.1 | |||
CX239635.1 | ||||
U66201.1 | AAB18917.1 |
More Information
Set Membership: |
Intragenic Exonic |