Assay Details
Target Gene Details
Entrez Gene ID: | 74374 |
Gene Name: | C-type lectin domain family 16, member A |
Gene Aliases: |
4932416N17Rik, curt, mKIAA0350 |
Location: |
Chr.16:10545339-10744878 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec16a | NM_001204229.1 | NP_001191158.1 | ||
NM_177562.5 | NP_808230.2 | |||
AK089327.1 | ||||
AK122255.1 | BAC65537.1 | |||
AK155314.1 | BAE33184.1 | |||
AK156927.1 | BAE33899.1 | |||
AK170754.1 | BAE42005.1 | |||
AK171134.1 | ||||
BC138907.1 | ||||
BC138909.1 |