Assay Details
Target Gene Details
Entrez Gene ID: | 12469 |
Gene Name: | chaperonin containing Tcp1, subunit 8 (theta) |
Gene Aliases: |
AI132397, Cctq, Tcpq |
Location: |
Chr.16:87483325-87495869 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cct8 | NM_009840.3 | 8 | 893 | NP_033970.3 |
AK010882.1 | 8 | 894 | BAB27244.1 | |
AK075711.1 | 8 | 874 | ||
AK075880.1 | 8 | 882 | BAC36025.1 | |
AK145661.1 | 8 | 877 | BAE26573.1 | |
AK145754.1 | 8 | 876 | BAE26629.1 | |
AK145918.1 | 8 | 895 | BAE26749.1 | |
AK147038.1 | 8 | 896 | BAE27627.1 | |
AK159969.1 | 8 | 885 | ||
AK168465.1 | 8 | 866 | ||
AK172867.1 | 8 | 864 | BAD32145.1 | |
AK191019.1 | 4 | 283 | ||
AK211232.1 | 4 | 283 | ||
AK214517.1 | 4 | 299 | ||
AK219341.1 | 4 | 283 | ||
BC009007.1 | 8 | 851 | AAH09007.1 | |
Z37164.1 | 8 | 897 | CAA85521.1 |
More Information
Set Membership: |
Intragenic Exonic |