Assay Details
Target Gene Details
Entrez Gene ID: | 12469 |
Gene Name: | chaperonin containing Tcp1, subunit 8 (theta) |
Gene Aliases: |
AI132397, Cctq, Tcpq |
Location: |
Chr.16:87483325-87495869 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cct8 | NM_009840.3 | 4 | 384 | NP_033970.3 |
AK010882.1 | 4 | 385 | BAB27244.1 | |
AK075711.1 | 4 | 365 | ||
AK075880.1 | 4 | 373 | BAC36025.1 | |
AK145661.1 | 4 | 368 | BAE26573.1 | |
AK145754.1 | 4 | 367 | BAE26629.1 | |
AK145918.1 | 4 | 386 | BAE26749.1 | |
AK147038.1 | 4 | 387 | BAE27627.1 | |
AK159969.1 | 4 | 376 | ||
AK168465.1 | 4 | 356 | ||
AK172867.1 | 4 | 355 | BAD32145.1 | |
AK182683.1 | 3 | 216 | ||
AK184650.1 | 3 | 184 | ||
AK189477.1 | 3 | 237 | ||
BC009007.1 | 4 | 342 | AAH09007.1 | |
Z37164.1 | 4 | 388 | CAA85521.1 |
More Information
Set Membership: |
Intragenic Exonic |