Assay Details
Target Gene Details
Entrez Gene ID: | 110213 |
Gene Name: | transmembrane BAX inhibitor motif containing 6 |
Gene Aliases: |
5031406P05Rik, Bi1, Tegt |
Location: |
Chr.15:99392947-99410049 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmbim6 | NM_001171034.1 | NP_001164505.1 | ||
NM_001171036.1 | NP_001164507.1 | |||
NM_026669.4 | NP_080945.1 | |||
AK019865.2 | BAB31892.1 | |||
AK036782.1 | BAC29575.1 | |||
AK036994.1 | BAC29662.1 | |||
AK049604.1 | BAC33837.1 | |||
AK050299.1 | BAC34174.1 | |||
AK050323.1 | BAC34188.1 | |||
AK088030.1 | BAC40107.1 | |||
AK088683.1 | BAC40503.1 | |||
AK139901.1 | BAE24176.1 | |||
AK159380.1 | BAE35035.1 | |||
AK166935.1 | BAE39128.1 | |||
AK217190.1 | ||||
BC005588.1 | AAH05588.1 | |||
BY272301.1 | ||||
CD554385.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |