Assay Details
Target Gene Details
Entrez Gene ID: | 22631 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta polypeptide |
Gene Aliases: |
1110013I11Rik, 14-3-3zeta, AI596267, AL022924, AU020854 |
Location: |
Chr.15:36770261-36802984 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ywhaz | NM_001253805.1 | NP_001240734.1 | ||
NM_001253806.1 | NP_001240735.1 | |||
NM_011740.3 | NP_035870.1 | |||
AK083368.1 | BAC38887.1 | |||
AK150381.1 | BAE29512.1 | |||
BB652039.1 | ||||
BC050891.1 | AAH50891.1 | |||
CF172373.1 | ||||
D87660.1 | BAA13421.1 |
More Information
Set Membership: |
Intragenic Non-exonic |