Assay Details
Target Gene Details
Entrez Gene ID: | 67367 |
Gene Name: | PAX3 and PAX7 binding protein 1 |
Gene Aliases: |
1810007M14Rik, C21orf66, Gcfc, Gcfc1, Pax3/7bp |
Location: |
Chr.16:91014037-91044734 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Paxbp1 | NM_026110.2 | 16 | 2448 | NP_080386.3 |
AK007365.1 | 10 | 1153 | BAB24988.2 | |
AK011477.1 | 6 | 532 | BAB27645.2 | |
AY033907.1 | 16 | 2233 | AAK68725.1 | |
AY033908.1 | 16 | 2250 | AAK68726.1 | |
BC014838.1 | 9 | 933 | ||
BC027145.1 | 8 | 868 | AAH27145.1 |
More Information
Set Membership: |
Intragenic Exonic |