Assay Details
Target Gene Details
Entrez Gene ID: | 268878 |
Gene Name: | ATPase type 13A5 |
Gene Aliases: |
C630015F21Rik |
Location: |
Chr.16:29231851-29378764 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp13a5 | NM_001284375.1 | NP_001271304.1 | ||
NM_175650.4 | NP_783581.2 | |||
AK083102.1 | ||||
AK083121.1 | BAC38770.1 | |||
AK158355.1 | BAE34470.1 | |||
BC115739.1 | ||||
CB524474.1 |