Assay Details
Target Gene Details
Entrez Gene ID: | 239857 |
Gene Name: | cell adhesion molecule 2 |
Gene Aliases: |
2900078E11Rik, 9330131D06, A830029E02Rik, Igdf4d, Igsf4d, NECL3, SynCAM 2, SynCAM2 |
Location: |
Chr.16:66655421-67621200 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cadm2 | NM_001145977.1 | 2 | 777 | NP_001139449.1 |
NM_178721.4 | 3 | 804 | NP_848836.2 | |
AK033973.1 | 2 | 404 | BAC28533.1 | |
AK038842.1 | 3 | 783 | BAC30148.1 | |
AK043760.1 | 3 | 787 | BAC31646.1 | |
AK046800.1 | 2 | 680 | BAC32876.1 | |
AK163636.1 | 3 | 781 | BAE37431.1 | |
AK165030.1 | 3 | 801 | BAE38008.1 | |
BC139401.1 | 3 | 373 | ||
BC139402.1 | 3 | 373 |