Assay Details
Target Gene Details
Entrez Gene ID: | 71726 |
Gene Name: | single-strand selective monofunctional uracil DNA glycosylase |
Gene Aliases: |
1200013B09Rik, A930006H09Rik, C85220 |
Location: |
Chr.15:103153290-103168895 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smug1 | AK004735.1 | BAB23517.1 | ||
AK020817.1 | ||||
AK045411.1 | ||||
AK090367.1 | ||||
AK172510.1 | BAE43042.1 | |||
AK198888.1 | ||||
AK204061.1 | ||||
BC050253.1 | 5 | 556 | AAH50253.1 |