Assay Details
Target Gene Details
Entrez Gene ID: | 30945 |
Gene Name: | ring finger protein 19A |
Gene Aliases: |
AA032313, Dorfin, Rnf19, UIP117, Ubce7ip2, XYbp |
Location: |
Chr.15:36239933-36283723 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnf19a | NM_013923.2 | NP_038951.1 | ||
AF120207.1 | AAF18303.1 | |||
AK082665.1 | ||||
AK147768.1 | BAE28125.1 | |||
AK190444.1 | ||||
BC040769.1 | AAH40769.1 | |||
DV656092.1 | ||||
X71642.1 | CAA50643.1 |
More Information
Set Membership: |
Intragenic Non-exonic |