Assay Details
Target Gene Details
Entrez Gene ID: | 56727 |
Gene Name: | myo-inositol oxygenase |
Gene Aliases: |
0610009I10Rik, AI314022, Aldrl6, C85427, RSOR |
Location: |
Chr.15:89334473-89337007 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Miox | NM_019977.2 | 4 | 322 | NP_064361.2 |
AF197127.1 | 4 | 320 | AAF25202.1 | |
AK002406.1 | 4 | 336 | ||
AK165481.1 | 1 | 1142 | ||
AY738257.1 | 3 | 292 | AAV65815.1 | |
BC013543.1 | 4 | 322 | AAH13543.1 |
More Information
Set Membership: |
Intragenic Exonic |