Assay Details
Target Gene Details
Entrez Gene ID: | 27401 |
Gene Name: | S-phase kinase-associated protein 2 (p45) |
Gene Aliases: |
4930500A04Rik, FBXL1, FWD1, p45 |
Location: |
Chr.15:9111982-9155425 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Skp2 | NM_001285980.1 | 4 | 655 | NP_001272909.1 |
NM_013787.3 | 4 | 725 | NP_038815.1 | |
AF083215.1 | 4 | 588 | AAD16037.1 | |
AK006221.1 | 4 | 655 | ||
AK032979.1 | 4 | 636 | BAC28108.1 | |
AK044212.1 | 4 | 626 | BAC31819.1 | |
AK166034.1 | 4 | 626 | BAE38535.1 | |
AK167677.1 | 4 | 623 | BAE39726.1 | |
BC092236.1 | 4 | 599 | AAH92236.1 | |
BY740905.1 |