Assay Details
Target Gene Details
Entrez Gene ID: | 14083 |
Gene Name: | PTK2 protein tyrosine kinase 2 |
Gene Aliases: |
FAK, FRNK, Fadk, mKIAA4203, p125FAK |
Location: |
Chr.15:73205102-73423275 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptk2 | NM_001130409.1 | 30 | 3175 | NP_001123881.1 |
NM_007982.2 | NP_032008.2 | |||
AB011499.1 | 30 | 2831 | BAC53890.1 | |
AB030035.1 | 27 | 2441 | BAC53924.1 | |
AK079821.1 | BAC37757.1 | |||
AK220543.1 | BAD90317.1 | |||
BU512211.1 | 1 | 86 | ||
BY297123.1 | 1 | 41 | ||
JN971016.1 | ||||
M95408.1 | AAA37592.1 |