Assay Details
Target Gene Details
Entrez Gene ID: | 73225 |
Gene Name: | family with sequence similarity 118, member A |
Gene Aliases: |
- |
Location: |
Chr.15:85037015-85116933 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam118a |
Target Gene Details
Entrez Gene ID: | 140557 |
Gene Name: | structural maintenance of chromosomes 1B |
Gene Aliases: |
SMC-1B, SMC1beta, Smc1l2 |
Location: |
Chr.15:85064689-85131957 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc1b | NM_080470.1 | NP_536718.1 | ||
AF303827.1 | AAL09333.1 | |||
BC129800.1 |