Assay Details
Target Gene Details
Entrez Gene ID: | 68501 |
Gene Name: | non-SMC element 2 homolog (MMS21, S. cerevisiae) |
Gene Aliases: |
1110014D18Rik, AI661537 |
Location: |
Chr.15:59374198-59601689 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nsmce2 | NM_001164604.1 | 1 | 104 | NP_001158076.1 |
NM_026746.3 | 1 | 104 | NP_081022.2 | |
AK003685.1 | 1 | 56 | BAB22938.1 | |
AK051277.1 | 1 | 56 | BAC34589.1 | |
AK188685.1 | ||||
AK211545.1 | ||||
AK212165.1 | ||||
BC009125.1 | AAH09125.1 | |||
DV057668.1 | 1 | 124 |