Assay Details
Target Gene Details
Entrez Gene ID: | 319944 |
Gene Name: | TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor |
Gene Aliases: |
150kDa, 4732460C16Rik, AI425886, CIF150, TAF2B, TAFII-150, TAFII150 |
Location: |
Chr.15:55015129-55072186 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Taf2 | NM_001081288.1 | 19 | 2696 | NP_001074757.1 |
AK147903.1 | 19 | 2698 | ||
BC147638.1 | 19 | 2647 | ||
BC151079.1 | 19 | 2647 |
More Information
Set Membership: |
Intragenic Exonic |