Assay Details
Target Gene Details
Entrez Gene ID: | 51812 |
Gene Name: | microspherule protein 1 |
Gene Aliases: |
C78274, ICP22BP, MSP58, P78 |
Location: |
Chr.15:99242817-99251961 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mcrs1 | NM_001164156.1 | NP_001157628.1 | ||
NM_016766.3 | NP_058046.2 | |||
AF015309.1 | AAC53590.1 | |||
AK088116.1 | BAC40155.1 | |||
AK158347.1 | BAE34465.1 | |||
AK158611.1 | ||||
AK167242.1 | BAE39364.1 | |||
AK187767.1 | ||||
BC003746.1 | AAH03746.1 | |||
BC085099.1 | AAH85099.1 | |||
BC108340.1 | AAI08341.1 |