Assay Details
Target Gene Details
Entrez Gene ID: | 20024 |
Gene Name: | SUB1 homolog (S. cerevisiae) |
Gene Aliases: |
AI842364, P15, P9, Pc4, Rpo2tc1 |
Location: |
Chr.15:11981338-11996031 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sub1 | NM_011294.3 | NP_035424.1 | ||
AK048371.1 | BAC33314.1 | |||
AK049260.1 | BAC33642.1 | |||
AK132021.1 | BAE20946.1 | |||
AK146335.1 | BAE27090.1 | |||
AK151734.1 | BAE30649.1 | |||
AK154114.1 | BAE32384.1 | |||
BC010967.1 | AAH10967.1 | |||
J03750.1 | AAA37317.1 |
More Information
Set Membership: |
Intragenic Non-exonic |