Assay Details
Target Gene Details
Entrez Gene ID: | 239510 |
Gene Name: | PHD finger protein 20-like 1 |
Gene Aliases: |
CGI-72, E130113K22Rik |
Location: |
Chr.15:66577517-66647043 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phf20l1 | NM_001081409.1 | 11 | 1534 | NP_001074878.1 |
AK032638.1 | 11 | 1549 | BAC27964.1 | |
AK040888.1 | 2 | 265 | BAC30733.1 | |
AK041825.1 | 1 | 632 | ||
AK080555.1 | 1 | 1167 | BAC37943.1 | |
AK206721.1 | 2 | 225 | ||
BC052212.1 | 10 | 1460 | AAH52212.1 | |
BC061492.1 | 10 | 1400 | AAH61492.1 | |
BC082777.1 | 11 | 1529 | AAH82777.1 | |
BC145689.1 | 10 | 1265 |