Assay Details
Target Gene Details
Entrez Gene ID: | 54615 |
Gene Name: | neuropeptide FF-amide peptide precursor |
Gene Aliases: |
- |
Location: |
Chr.15:102523839-102524942 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npff | NM_018787.1 | NP_061257.1 | ||
NR_033409.1 | 1 | 702 | ||
AF148701.1 | AAD39829.1 | |||
BC107294.1 | 1 | 376 | AAI07295.1 | |
BC107295.1 | 1 | 381 | AAI07296.1 |