Assay Details
Target Gene Details
Entrez Gene ID: | 20788 |
Gene Name: | sterol regulatory element binding factor 2 |
Gene Aliases: |
AI608257, SREBP-2, SREBP2, SREBP2gc, bHLHd2, lop13, nuc |
Location: |
Chr.15:82147269-82205379 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srebf2 | NM_033218.1 | 17 | 3054 | NP_150087.1 |
AF374267.1 | 17 | 2907 | AAK54763.1 | |
AK155857.1 | 18 | 3002 | BAE33463.1 | |
AK172219.1 | 17 | 3063 | ||
CB228109.2 | 2 | 254 |