Assay Details
Target Gene Details
Entrez Gene ID: | 20788 |
Gene Name: | sterol regulatory element binding factor 2 |
Gene Aliases: |
AI608257, SREBP-2, SREBP2, SREBP2gc, bHLHd2, lop13, nuc |
Location: |
Chr.15:82147269-82205379 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srebf2 | NM_033218.1 | 15 | 2773 | NP_150087.1 |
AF374267.1 | 15 | 2626 | AAK54763.1 | |
AK155857.1 | 16 | 2721 | BAE33463.1 | |
AK172219.1 | 15 | 2781 | ||
BQ963055.1 | 6 | 832 |