Assay Details
Target Gene Details
Entrez Gene ID: | 77980 |
Gene Name: | SET binding factor 1 |
Gene Aliases: |
2610510A08Rik, B230113C15Rik, Mtmr5, mKIAA3020 |
Location: |
Chr.15:89288236-89315311 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sbf1 | NM_001081030.2 | 20 | 2717 | NP_001074499.2 |
NM_001170561.1 | 20 | 2717 | NP_001164032.1 | |
AK129485.1 | 20 | 2661 | BAC98295.1 | |
AK147332.1 | 20 | 2674 | ||
BC098209.1 | 1 | 60 | AAH98209.1 | |
BC157935.1 | 20 | 2607 | ||
BC158013.1 | 20 | 2607 | ||
BC172094.1 | 20 | 2607 |
More Information
Set Membership: |
Intragenic Exonic |