Assay Details
Target Gene Details
Entrez Gene ID: | 67747 |
Gene Name: | RIB43A domain with coiled-coils 2 |
Gene Aliases: |
4930579A10Rik, Trib |
Location: |
Chr.15:85132060-85144569 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ribc2 | NM_026357.2 | NP_080633.2 | ||
AK006801.1 | ||||
AK016311.1 | BAB30190.2 | |||
AK017028.1 | ||||
BC112373.1 | AAI12374.1 | |||
BY716712.1 |
More Information
Set Membership: |
Intragenic Non-exonic |