Assay Details
Target Gene Details
Entrez Gene ID: | 105787 |
Gene Name: | protein kinase, AMP-activated, alpha 1 catalytic subunit |
Gene Aliases: |
AI194361, AI450832, AL024255, AMPKalpha1, C130083N04Rik |
Location: |
Chr.15:5143861-5181899 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prkaa1 | NM_001013367.3 | NP_001013385.3 | ||
AK147811.1 | ||||
AK160612.1 | BAE35914.1 | |||
AY885266.1 | AAW79567.1 | |||
BC086695.1 |
More Information
Set Membership: |
Intragenic Non-exonic |