Assay Details
Target Gene Details
Entrez Gene ID: | 53357 |
Gene Name: | phospholipase A2, group VI |
Gene Aliases: |
BB112799, PNPLA9, iPLA(2)beta, iPLA2, iPLA2beta |
Location: |
Chr.15:79286228-79328530 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g6 | NM_001199023.1 | 4 | 1253 | NP_001185952.1 |
NM_001199024.1 | 4 | 1253 | NP_001185953.1 | |
NM_001199025.1 | 5 | 758 | NP_001185954.1 | |
NM_016915.4 | 4 | 799 | NP_058611.1 | |
AF259401.1 | 3 | 533 | AAF72651.1 | |
AK144495.1 | 5 | 632 | BAE25917.1 | |
AK153333.1 | 4 | 1059 | BAE31912.1 | |
BC003487.1 | 4 | 592 | AAH03487.1 | |
BC052845.1 | 4 | 624 | AAH52845.1 | |
BC057209.1 | 4 | 846 | AAH57209.1 | |
CF531790.1 | 4 | 682 | ||
U88624.2 | 3 | 475 | AAB48511.2 |
More Information
Set Membership: |
Intragenic Exonic |