Assay Details
Target Gene Details
Entrez Gene ID: | 53357 |
Gene Name: | phospholipase A2, group VI |
Gene Aliases: |
BB112799, PNPLA9, iPLA(2)beta, iPLA2, iPLA2beta |
Location: |
Chr.15:79286228-79328530 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g6 | NM_001199023.1 | 15 | 2859 | NP_001185952.1 |
NM_001199024.1 | 14 | 2694 | NP_001185953.1 | |
NM_001199025.1 | 15 | 2199 | NP_001185954.1 | |
NM_016915.4 | 14 | 2240 | NP_058611.1 | |
AF259401.1 | 14 | 2139 | AAF72651.1 | |
AK144495.1 | 15 | 2073 | BAE25917.1 | |
AK153333.1 | 14 | 2500 | BAE31912.1 | |
BC003487.1 | 14 | 2033 | AAH03487.1 | |
BC052845.1 | 14 | 2065 | AAH52845.1 | |
BC057209.1 | 15 | 2452 | AAH57209.1 | |
U88624.2 | 13 | 1916 | AAB48511.2 |
More Information
Set Membership: |
Intragenic Exonic |