Assay Details
Target Gene Details
Entrez Gene ID: | 70363 |
Gene Name: | family with sequence similarity 135, member B |
Gene Aliases: |
1700010C24Rik, A830008O07Rik |
Location: |
Chr.15:71438951-71728436 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam135b | NM_177819.3 | NP_808487.2 | ||
AK005816.1 | BAB24252.2 | |||
BC119174.1 | ||||
BC137579.1 |