Assay Details
Target Gene Details
Entrez Gene ID: | 23880 |
Gene Name: | FYN binding protein |
Gene Aliases: |
ADAP, B630013F22Rik, FYB-120/130, Slap130, p120/130, p120/p130 |
Location: |
Chr.15:6522797-6665608 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fyb | NM_001278269.1 | 7 | 1661 | NP_001265198.1 |
NM_011815.5 | 7 | 1661 | NP_035945.1 | |
AF001863.1 | 7 | 1637 | AAB62227.1 | |
AF061744.1 | 7 | 1637 | AAD03267.1 | |
AK038225.1 | 7 | 1689 | ||
AK137907.1 | 7 | 1617 | ||
BC141352.1 | 7 | 1602 |
More Information
Set Membership: |
Intragenic Exonic |