Assay Details
Target Gene Details
Entrez Gene ID: | 97998 |
Gene Name: | DEP domain containing MTOR-interacting protein |
Gene Aliases: |
Depdc6 |
Location: |
Chr.15:55112317-55259273 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Deptor | NM_001037937.3 | NP_001033026.1 | ||
NM_145470.3 | NP_663445.2 | |||
AK139888.1 | BAE24172.1 | |||
AK145208.1 | BAE26298.1 | |||
AK188097.1 | ||||
AK220300.1 | BAD90225.1 |
More Information
Set Membership: |
Intragenic Non-exonic |